The technical specifications for Model Number 987 are as follows:
- Filter Type: HEPA Filter, Adsorption Module
- Contaminants Removed: VOCs, odors, 0.3 microns, viruses, bacteria, smoke, fumes, fungus, mold, pollen, mites, dust, fibers
- Weight: 17 lbs.
- Dimensions: HEPA - 12" x 12" x 6"; Adsorption Module - 12" x 12" x 2"
- Filter Efficiency: HEPA - 99.97% at 0.3 microns
- Shipping box dimensions: 14" x 14" x 10"
おすすめの製品
法的情報
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
最新バージョンのいずれかを選択してください:
-
What is the exterior size of the Z687308 filter?
1 回答-