Merck
Z250325
Sigma-Aldrich
Aldrich® azeotropic distillation apparatus
joint: threaded 14/20
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥133,000
¥133,000
Check Cart for Availability
サイズを選択してください
表示を変更する
1 EA
¥133,000
About This Item
UNSPSCコード:
41104812
NACRES:
NB.43
¥133,000
Check Cart for Availability
ジョイント
joint: threaded14/20
詳細
All connections are made using greaseless, 14/20 screw-thread joints. All parts are interchangeable with those contained in the Aldrich All-In-One glassware kit, Z166898. Includes distillation head and 100 mL round-bottom flask. Dropping funnel sold separately, Z167541. Effective for the removal of water from condensation reactions[1].
Azeotropic Technique
1.
The solvent/H2O azeotrope is cooled completely by a water-cooled condenser and directed to the extraction reservoir (filled with 4Å molecular sieves through a 14/20 joint). The reservoir joint* can also be used as a septum-inlet for the removal of water via syringe.
2. The solvent and any volatile reagents are recycled through the sieves at ambient temperature.
3. Additional reagent can be added to the reservoir (via dropping funnel connected at the reservoir joint) as a means of slow addition to a mixture at reflux.
*Use with rubber septa Z564702, Z564699, Z512125, Z512117
関連事項
ねじ込み式ジョイントの扱い方の詳細については、Threaded Ground Glass Tutorialをご参照ください。
法的情報
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
最新バージョンのいずれかを選択してください:
試験成績書(COA)
文書ライブラリにアクセスする
Barta, N.S., Paulvannan, K., Schwarz, J.B., Stille, J.R.
Synthetic Communications, 24, 583-583 (1994)