SYNF904500G
別名:
Synthware F904500G, modified Schlenk reaction flask
SYNF904500G-1EA
¥15,880
1 of 10
SYNF904000G
Synthware™ pear-shaped Schlenk flask with stopcock
Sigma-Aldrich
Z515981
Schlenk reaction and storage tube
Supelco
1.00306
Hydrobromic acid 47%
722472
1-ブロモ-4-(ジフルオロメチル)ベンゼン
Z722642
AlumaSeal® CS Films for cold storage
70956
1-ブチル-3-メチルイミダゾリウム ヘキサフルオロホスファート
Z516023
Z567183
Ace丸底圧力フラスコ
Z530964
Aldrich® Liebig condenser
SYNF10401LA
Synthware™ 47 mm filtration apparatus with stainless steel support, complete
borosilicate glass flaskpear-shaped single-neck
joint: ST/NS24/40
pkg of 1 ea
Synthware F904500G
500 mL
類似した製品をお探しですか? 訪問 製品比較ガイド
最新バージョンのいずれかを選択してください:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 資料 section.