Merck
BMSV0020
Sigma-Aldrich
Aspire Laboratory Aspirator
includes base with internal vacuum pump, 2 L Polycarbonate bottle with lid, silicone tubing, handheld vacuum controller and single channel adapters, AC/DC input 115 V AC, US 3-pin plug
別名:
aspiration system, vacuum aspirator
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥180,400
¥180,400
Check Cart for Availability
サイズを選択してください
表示を変更する
1 EA
¥180,400
About This Item
UNSPSCコード:
41161502
¥180,400
Check Cart for Availability
メーカー/製品名
Benchmark Scientific V0020
パラメーター
-0 mbar vacuum (-750 mbar)
外寸(幅×奥行×高さ)
9.5 in. × 7.9 in. × 13.9 in.
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The ASPIRE Laboratory Aspirator incorporates a quiet, maintenance-free pump inside a sturdy, yet compact base. The vacuum is fully adjustable, with a pressure gauge that displays the vacuum level, allowing full control of aspirating speed. The ASPIRE is an ideal choice for a wide range of laboratory aspirating applications, including removal of media from tissue culture vessels or plates, and supernatants.
法的情報
Aspire is a trademark of Benchmark Scientific, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)