Documenti
Z761001
TPP® easy read cell counter
Sinonimo/i:
Automated cell counter, Cell counting device
About This Item
Codice UNSPSC:
42181504
NACRES:
NB.22
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
TPP 87010
Campo di misura
0-5 μL
Descrizione generale
Features of the "easy read" mesuring device:
  • Reading device with magnifying glass and cross line
  • Optimal usage in combination with the PCV tube
  • Reproducible cell pellet measuring possible
  • Reusable product, no further investments necessary
  • Measuring range : 0 - 5 μl
Note legali
TPP is a registered trademark of TPP Techno Plastic Products AG