Documenti fondamentali
Z355453
Millipore
Millipore® Sterifil® filtration system
Complete System
Scegli un formato
1 EA
226,00 €
226,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
226,00 €
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.24
226,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polypropylene housing
polysulfone housing
Sterilità
sterile
Caratteristiche
cap plug seal
Confezionamento
pack of 1
Produttore/marchio commerciale
Millipore XX1104700
Descrizione generale
A reusable, aseptic filter system that can be autoclaved with the filter of choice in place. Construction consists of a polysulfone funnel, funnel cover, and cover; polypropylene base and filter support screen; silicone stopper and O-ring. Accepts a 47 mm filter. Capacity is 250 mL.
Perforated stopper on holder outlet mounts in a standard 1-liter filtering flask or Millipore filtering manifold. Two ports on receiver flask accept either 6 mm (1/4 in.) I.D. tubing or male luer-slip connection for vacuum and drain or vent.
Four female luer-slip ports on funnel cover can be used for vent and aseptic inlet.
Includes polysulfone funnel, funnel cover, receiver flask and cover; polypropylene holder base, and filter support screen; silicone stopper
Note legali
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Sterifil is a registered trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)