TMO436111
Nunc® 96-well immuno plate
polystyrene, non-sterile, without lid, white, case of 80 ea
Sinonimo/i:
assay microplate, surface treated microplate
About This Item
Codice UNSPSC:
41122102
NACRES:
NB.15
Prodotti consigliati
Materiali
polystyrene
white
Sterilità
non-sterile
Confezionamento
case of 80 ea
Produttore/marchio commerciale
Nunc 436111
Categorie correlate
Descrizione generale
Thermo Scientific White 96-Well Immuno Plates are used to perform both quantitative and qualitative immunoassays, such as ELISA and binding assays. It is designed to be compatible with commonly used instruments. These plates ensure excellent performance, consistent results between different lots, and reproducibility across individual wells.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)