HS15994P
Premium slide box
100 place, foam, red plastic
Sinonimo/i:
Schiebekasten, slide box microscope, slide storage, slide storage box
Scegli un formato
25,10 €
Scegli un formato
About This Item
25,10 €
Prodotti consigliati
Materiali
nickel closure
red plastic
Caratteristiche
Holds 100 x slides 76x26 mm
Produttore/marchio commerciale
Heathrow Scientific HS15994P
Lungh. × Largh. × Alt.
208 × 175 × 34 mm
Categorie correlate
Caratteristiche e vantaggi
- Molded, rigid boxes securely hold slides in grooved slots
- Durable ABS plastic boxes are available with cork lining only
- Both the lining and the slots are numbered to correspond to the slide inventory sheet
- Feature thumb-latch lock mechanism
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)