Documenti fondamentali
GBL620005
Grace Bio-Labs SecureSeal adhesive sheets
thickness 0.60 mm, Double Sided, L × W 7 in. × 10 in.
Sinonimo/i:
silicone sheet with double sided adhesive
Scegli un formato
1 EA
187,00 €
187,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
187,00 €
About This Item
Codice UNSPSC:
31201600
NACRES:
NB.17
187,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Sterilità
non-sterile
Caratteristiche
Double Sided
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Grace Bio-Labs 620005
Lungh. × largh.
7 in. × 10 in.
Spessore
0.60 mm
Descrizione generale
Grace Bio-Labs SecureSeal adhesive sheets are made up of SecureSeal adhesive, which is used to make HybriWell and SecureSeal incubation chambers. It has a thin, double-sided tape on a non-woven carrier, a clear, polyester liner on one surface, and a white clay-coated paper on the other. SecureSeal sheets are available in various thicknesses.
These adhesive sheets are made using the same SecureSeal adhesive as is used to make HybriWell and SecureSeal Incubation Chambers. Thin, double-sided tape on a non-woven carrier. Clear, polyester liner on one surface, white clay coated paper on the other. SecureSeal Sheets consist of various thickness to choose from. These double sided adhesive sheets measure 7 x 10 inches.
Note legali
HybriWell is a trademark of Grace Bio-Labs, Inc.
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)