Documenti fondamentali
BAF134880075
Bel-Art® Write-on Label Tape
roll size 40 yard, orange, width 3/4 in., pk of 4
Sinonimo/i:
labelling tape, moisture resistant label tape, solvent resistant label tape
Scegli un formato
4 EA
76,90 €
76,90 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
4 EA
76,90 €
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
76,90 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
orange
Confezionamento
pk of 4
Produttore/marchio commerciale
Bel-Art F13488-0075
Dimensioni rotolo
40 yard
Larghezza
19.05 mm
3/4 in.
Descrizione generale
Select from six tape colors that correspond with familiar hazard color codes to highlight safety information written on the labels. Use yellow for chemical reactivity, green for safety labels, blue indicates health hazards, red alerts to flammability; orange indicates a biohazard and white can describe a specific hazard. The tape is custom manufactured from proprietary materials that resist moisture and most solvents. Will adhere to PTFE fluoropolymer resin covered materials and any clean surface. A maximum temperature of 247 degrees C for 30 minutes can be endured.
Caratteristiche e vantaggi
  • No sticky residue lingers when tape is stripped off
  • Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)