Documenti
Z742377
Sigma-Aldrich
CANgas inner adaptor
About This Item
Codice UNSPSC:
12142100
NACRES:
NA.22
Livello qualitativo
Descrizione generale
1/8′′ NPT inner adaptor for pressure can. Applicable for Messer® CANgas
Altre note
Product is currently only available to our customers in Europe
Note legali
Messer is a registered trademark of Messer Group GmbH
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Air synthetic, Messer® CANgas, 99.999%
Sigma-Aldrich
769096
Air
Carbon dioxide Messer® CANgas, 99.995%
Sigma-Aldrich
769002
Carbon dioxide
Propane 99.95%, Messer® CANGas
Sigma-Aldrich
769037
Propane
Helium Messer® CANGas, 99.999%
Sigma-Aldrich
768995
Helium
Xenon 99.99%, Messer® CANGas
Sigma-Aldrich
769118
Xenon
Ethylene Messer® CANGas, 99.95%
Sigma-Aldrich
768987
Ethylene