Documenti fondamentali
Z742064
Sigma-Aldrich
KitAlysis Benchtop Inertion Box
Sinonimo/i:
Benchtop Inertion Box
Scegli un formato
1 EA
979,00 €
979,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Richiedi un ordine bulk
Scegli un formato
Cambia visualizzazione
1 EA
979,00 €
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
979,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Richiedi un ordine bulk
Descrizione generale
Provides an inert environment to run oxygen sensitive cross-coupling reactions in a laboratory fume hood.
Designed to be used with KitAlysis High-Throughput Screening Kits and KitAlysis 24-Well Reaction Block.
Caratteristiche e vantaggi
  • Run 24 micro-scale reactions in parallel under an inert atmosphere.
  • Provides a constant flow of inert gas over working area.
  • Easy attachment of tubing (not included) from inert gas valve on hood or stand-alone tank.
Note legali
KitAlysis is a trademark of Sigma-Aldrich Co. LLC
Comunemente ordinati con questo prodotto
N° Catalogo
Descrizione
Determinazione del prezzo
Raccomandato
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Articoli
Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.
KitAlysis High-Throughput Screening Kits provide solution to efficiently identify or optimize suitable catalytic reaction conditions. Chemist can rapidly run 24 unique micro scale reactions in parallel with tailored conditions.
PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.
Materials Included in your KITALYSIS-24PD-2PK High-Throughput Screening Kit
Protocolli
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.
Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit
A step-by-step protocol guide for KitAlysis Suzuki-Miyaura Cross-Coupling Reaction Screening Kit.
Vedi tutto
Contenuto correlato
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.