Documenti fondamentali
Z678287
Finnpipette® F2 variable volume pipette
volume 0.5-5 μL , Micro-tip
Scegli un formato
1 EA
384,00 €
384,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
384,00 €
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
384,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Caratteristiche
color-coded pink
electronic: no
single-channel
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Finn 4642020
Volume
0.5-5 μL , Micro-tip
Descrizione generale
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50μl and below
Note legali
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)