The product dimensions are 575 mm x 400 mm x 12.5 mm (23 x 16 x 1/2 in). The board is made of HDPE which is not autoclavable but may be sanitized using a 70% ethanol wash.
Z675776
DissecTable™ reversible dissecting board
polyethylene, pkg of 1 ea
Scegli un formato
364,00 €
Scegli un formato
About This Item
364,00 €
Prodotti consigliati
Materiali
polyethylene
Confezionamento
pkg of 1 ea
Descrizione generale
One side has a large cutting area including dimensional scales in inches and centimeters, along with a 60 x 80 mm grid made of 48 x 10 mm squares. Six dimensional circles are also printed from 1/8 to 5/8 in. and 4 mm to 14 mm in diameter. The other side offers 2 cutting boards half the size with the same dimensional features printed on each one. All corners have rubber feet giving more stability to the working surface.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
-
LS, What are the dimensions of this board and is it possible to autoclave this item multiple times? with regards
1 risposta-