Z551864
Aldrich® check-valve bubbler with SafetyBarb® hose connectors
single inlet tube, top outlet
Sinonimo/i:
bubblers, scientific bubbler
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Capacità
15 mL
Descrizione generale
Check-valve ball prevents oil from being drawn into the system. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)