Documenti
Z510351
Sigma-Aldrich
Vigreux distilling column
bottom joint: ST/NS 14/20
About This Item
Codice UNSPSC:
40161600
NACRES:
NB.43
Giunto
joint: ST/NS bottom14/20
Categorie correlate
Descrizione generale
Vigreux indents run the entire length of the column and act as the packing for the column.