Documenti fondamentali
Z506559
Perfektum® one-way stainless steel stopcock
female Luer to male Luer lock
Scegli un formato
1 EA
341,00 €
341,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
341,00 €
About This Item
Codice UNSPSC:
41122000
NACRES:
NB.03
341,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Caratteristiche
CE compliant
Produttore/marchio commerciale
Cadence Science 6021SS
Parametri
45-60 psi pressure range (this fitting is pressure tested using water)
Descrizione generale
Made from corrosion-resistant 303 stainless steel.
These adapters have been specifically designed to minimize stripping of plastic Luers when mated with metal Luers.
Manufactured to ISO 594 1/2 standards and meet FDA and CE requirements.
Note legali
Perfektum is a registered trademark of Cadence Science
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 8
1 of 8
Luer-to-Closed end stopper male Luer lock to closed end
Z117951
Luer-to-Closed end stopper
3-way stopcock female Luer lock to male Luer lock, right side, female Luer lock, compression-nut
CAD6011
3-way stopcock
3-way stopcock female Luer lock to female Luer lock, male Luer lock, in. Y in. type
CAD6056
3-way stopcock
3-way stopcock female Luer lock to male Luer lock, left side, female Luer lock
CAD6017
3-way stopcock
3-way stopcock female Luer lock to female Luer lock, right side, female Luer lock
CAD6002
3-way stopcock
3-way stopcock female Luer lock to male Luer, right side, female Luer lock, compression-nut
CAD6001
3-way stopcock