Key Documents
Z407380
Chemical Information Database
About This Item
Codice UNSPSC:
41100000
Descrizione generale
Information for tens of thousands of compounds in a single source. The Windows-based full-text retrieval system gives users information on many commonly used compounds. Data have been compiled from many secondary sources, including NIOSH Pocket Guide to Chemical Hazards, NJ Hazardous Substances Fact Sheets, TSCA Chemical Inventory, EPA Chemical Fact Sheets and Chemical Summary Sheets, EPA′s IRIS, CDC′s listing of hazardous substances, CDC Agency for Toxic Substances and Disease Registry (ATSDR) fact sheets, OSHA Chemical Sampling Information Sheets, and others.
Requisiti del sistema
Windows 3.1 or later
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 5
1 of 5
Acetone HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
650501
Acetone
2-Propanolo ≥99.7%, FCC, FG
Sigma-Aldrich
W292907
2-Propanolo
Caffeina powder, ReagentPlus®
Sigma-Aldrich
C0750
Caffeina