Documenti
Z303941
Clips for Quickfit® flat-flange reaction flask lids
For 100 mm bore
Sinonimo/i:
JC100F
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
stainless steel
Etichetta
D
Sterilità
non-sterile
Confezionamento
pack of 1
Produttore/marchio commerciale
Quickfit JC 100F
Disponibilità
not available in USA and Canada
Descrizione generale
Designed for securing flat flange joints, these clips have a hinged ′spring wire′ design that tightens using a wing nut.
Note legali
Quickfit is a registered trademark of SciLabware Limited