Documenti fondamentali
Z146218
Supelco
Aldrich® rectangular TLC developing tanks, lid only
For Z126195
About This Item
Codice UNSPSC:
41115711
NACRES:
NC.07
tecniche
thin layer chromatography (TLC): suitable
Descrizione generale
TLC developing tanks are used to run TLC plates for separation. It is important to maintain saturated conditions during the analysis which can be achieved by using small tanks, filter paper liners and sufficient solvents. The tank can be equilibrated for 30 mins prior to running the plate to yield good results.[1] Lid is used to cover the tank.
Applicazioni
Aldrich® rectangular TLC developing tanks, lid only may be used to develop TLC plates.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
An overview on thin layer chromatography
Bele, Archana A., and Anubha Khale.
International Journal of Pharmaceutical Sciences and Research, 2, 256-267 (2011)