Documenti fondamentali
THS02134
ThalesNano CatCart® catalyst cartridge system, 70 mm L
5% Pd/CaCO3, poisoned with lead
Sinonimo/i:
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
Codice UNSPSC:
60104500
NACRES:
NB.51
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
ThalesNano THS 02134
Diametro
4 mm
Descrizione generale
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Note legali
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)