SYNF100002
Synthware™ 47 mm filtration apparatus clamp
Sinonimo/i:
Synthware™ 47 mm filtration apparatus with fritted support
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F100002
Categorie correlate
Descrizione generale
Using 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks. Support base has a sealed-in coarse porosity fritted disc. Complete assembly includes a 300mL graduated funnel, aluminum clamp, support base and l liter filtering flask.
Note legali
Synthware is a trademark of Kemtech America Inc.