DWKW217931
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity (1,250 mL), white polypropylene cap, (PTFE faced LDPE liner), case of 6 ea
Sinonimo/i:
W217931
Scegli un formato
60,70 €
Scegli un formato
About This Item
60,70 €
Prodotti consigliati
Materiali
(PTFE faced LDPE liner)
amber soda-lime glass bottle
white polypropylene cap
agenzia
suitable for EPA 1613
Sterilità
non-sterile
Caratteristiche
with cap
with cap
Confezionamento
case of 6 ea
Produttore/marchio commerciale
WHEATON® W217931
Diametro bottiglia × alt.
(108 mm) (192 mm)
Capacità
(1,250 mL)
Raccordi
70-400 neck
Caratteristiche e vantaggi
- Ideal for storage of light sensitive samples or dry powders
- Easy removal of contents
- HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification
- Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
- Amber, USP Type III soda-lime glass
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Contenuto correlato
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.