Documenti
CLS5400250
Pyrex® iodine determination flask, with Pyrex® ST stopper
capacity 250 mL
Sinonimo/i:
flask erlenmeyer, flasks erlenmeyer, glass flasks, Corning® 5400
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
wide-mouth
Giunto
joint ST/NS stopper22
Sterilità
non-sterile
Caratteristiche
cap: no
closure type ST stopper
Confezionamento
pack of 1
case of 8
Produttore/marchio commerciale
Corning 5400-250
Capacità
250 mL
Descrizione generale
Flasks are blown in specillay designed molds, thereby insuring uniformity and proper contour. The stopper projects above the liquid seal trough to facilitate removal.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.