Documenti
BR759241
BRAND® cap for UV-cuvette micro
yellow
Sinonimo/i:
Accessories, Caps, Cuvettes, UV-Cuvettes
About This Item
Codice UNSPSC:
41121800
NACRES:
SB.32
Materiali
PE
polyethylene
yellow
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
BRAND 759241
Descrizione generale
Round covers allow a tight closure and make it possible to store samples down to -20 °C.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG