Documenti fondamentali
BR704272
BRAND® table/shelf clamp for remote dispensing Dispensette®
Sinonimo/i:
Accessories, Bottle-top Dispenser, Dispenser, Dispensette®
Scegli un formato
1 EA
18,50 €
18,50 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
18,50 €
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.01
18,50 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 704272
Categorie correlate
Descrizione generale
Dispense accurate volumes directly from drums and bulk refills.The Dispensette® can be mounted on a wall, a ring stand or on lab furniture.A filter in the drum adapter minimizes risk of contaminating highpurity reagents when air is drawn in for pressure compensation after dispensing.A quick-release connector with integrated valves allows quick changing of the bulk container.The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette®.The max. delivery height is approximately 1.2 m.Operating Exclusions: Never use the remote dispensing system: 1. with recirculation valve. It has to be removed before use! 2. for pressurized vessels 3. for liquids attacking borosilicate glass, Al2O3-ceramic, PFA, ETFE, FEP or PTFE 4. for Peroxide (due to catalytic reaction) 5. for carbon disulfide (CS2), due to risk of explosion!
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)