- Antibiotics do not affect the degradation of fungicides and enhance the mineralization of chlorpyrifos in biomixtures.
Antibiotics do not affect the degradation of fungicides and enhance the mineralization of chlorpyrifos in biomixtures.
Ecotoxicology and environmental safety (2017-02-20)
Humberto Castillo-González, Marta Pérez-Villanueva, Mario Masís-Mora, Víctor Castro-Gutiérrez, Carlos E Rodríguez-Rodríguez
PMID28214645
KIVONAT
The use of antibiotics in agriculture produces residues in wastewaters. The disposal of such wastewaters in biopurification systems (BPS) employed for the treatment of pesticides could result in the inhibition of the degrading capacity of the biomixtures used in the BPS. We assayed the effect of two commercial formulations of antibiotics used in agriculture, one containing kasugamycin (KSG) and the other oxytetracycline plus gentamicin (OTC+GTM), on the biomixture performance. Doses from 0.1mgkg
ANYAGOK