Ugrás a tartalomra
Merck
  • CTNNB1 in mesenchyme regulates epithelial cell differentiation during Müllerian duct and postnatal uterine development.
CTNNB1 in mesenchyme regulates epithelial cell differentiation during Müllerian duct and postnatal uterine development.
Molecular endocrinology (Baltimore, Md.) (2013-08-02)
C Allison Stewart, Ying Wang, Margarita Bonilla-Claudio, James F Martin, Gabriel Gonzalez, Makoto M Taketo, Richard R Behringer
KIVONAT
Müllerian duct differentiation and development into the female reproductive tract is essential for fertility, but mechanisms regulating these processes are poorly understood. WNT signaling is critical for proper development of the female reproductive tract as evident by the phenotypes of Wnt4, Wnt5a, Wnt7a, and β-catenin (Ctnnb1) mutant mice. Here we extend these findings by determining the effects of constitutive CTNNB1 activation within the mesenchyme of the developing Müllerian duct and its differentiated derivatives. This was accomplished by crossing Amhr2-Cre knock-in mice with Ctnnb1 exon (ex) 3(f/f) mice. Amhr2-Cre(Δ/+); Ctnnb1 ex3(f/+) females did not form an oviduct, had smaller uteri, endometrial gland defects, and were infertile. At the cellular level, stabilization of CTNNB1 in the mesenchyme caused alterations within the epithelium, including less proliferation, delayed uterine gland formation, and induction of an epithelial-mesenchymal transition (EMT) event. This EMT event is observed before birth and is complete within 5 days after birth. Misexpression of estrogen receptor α in the epithelia correlated with the EMT before birth, but not after. These studies indicate that regulated CTNNB1 in mesenchyme is important for epithelial cell differentiation during female reproductive tract development.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone LN-6, ascites fluid