Ugrás a tartalomra
Merck
  • Palladium-catalyzed cross-coupling of stereospecific potassium cyclopropyl trifluoroborates with aryl bromides.
Palladium-catalyzed cross-coupling of stereospecific potassium cyclopropyl trifluoroborates with aryl bromides.
Organic letters (2004-01-30)
Guo-Hua Fang, Zheng-Jun Yan, Min-Zhi Deng
KIVONAT
[reaction: see text] Stereospecific cyclopropanation of alkenylboronic esters of pinacol followed by in situ treatment with excess KHF(2) afforded the corresponding potassium cyclopropyl trifluoroborates in high yields, which then underwent Suzuki-Miyaura cross-coupling reactions with aryl bromides to give cyclopropyl-substituted arenes in good yields with retention of configuration. This promises to be a useful method for the synthesis of enantiomerically pure cyclopropanes.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Potassium cyclopropyltrifluoroborate, ≥99%