Ugrás a tartalomra
Merck
  • Confirmation of and explanations for elevated blood lead and other disorders in children exposed to water disinfection and fluoridation chemicals.
Confirmation of and explanations for elevated blood lead and other disorders in children exposed to water disinfection and fluoridation chemicals.
Neurotoxicology (2007-04-11)
Myron J Coplan, Steven C Patch, Roger D Masters, Marcia S Bachman
KIVONAT
Silicofluorides (SiFs), fluosilicic acid (FSA) and sodium fluosilicate (NaFSA), are used to fluoridate over 90% of US fluoridated municipal water supplies. Living in communities with silicofluoride treated water (SiFW) is associated with two neurotoxic effects: (1) Prevalence of children with elevated blood lead (PbB>10microg/dL) is about double that in non-fluoridated communities (Risk Ratio 2, chi2p<0.01). SiFW is associated with serious corrosion of lead-bearing brass plumbing, producing elevated water lead (PbW) at the faucet. New data refute the long-prevailing belief that PbW contributes little to children's blood lead (PbB), it is likely to contribute 50% or more. (2) SiFW has been shown to interfere with cholinergic function. Unlike the fully ionized state of fluoride (F-) in water treated with sodium fluoride (NaFW), the SiF anion, [SiF6]2- in SiFW releases F- in a complicated dissociation process. Small amounts of incompletely dissociated [SiF6]2- or low molecular weight (LMW) silicic acid (SA) oligomers may remain in SiFW. A German PhD study found that SiFW is a more powerful inhibitor of acetylcholinesterase (AChE) than NaFW. It is proposed here that SiFW induces protein mis-folding via a mechanism that would affect polypeptides in general, and explain dental fluorosis, a tooth enamel defect that is not merely "cosmetic" but a "canary in the mine" foretelling other adverse, albeit subtle, health and behavioral effects. Efforts to refute evidence of such effects are analyzed and rebutted. In 1999 and 2000, senior EPA personnel admitted they knew of no health effects studies of SiFs. In 2002 SiFs were nominated for NTP animal testing. In 2006 an NRC Fluoride Study Committee recommended such studies. It is not known at this writing whether any had begun.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Fluosilicic acid, purum, 33.5-35%