Ugrás a tartalomra
Merck
  • Transcriptome profiling of rabbit parthenogenetic blastocysts developed under in vivo conditions.
Transcriptome profiling of rabbit parthenogenetic blastocysts developed under in vivo conditions.
PloS one (2012-12-20)
Carmen Naturil-Alfonso, María Dels Desamparats Saenz-de-Juano, David S Peñaranda, José S Vicente, Francisco Marco-Jiménez
KIVONAT
Parthenogenetic embryos are one attractive alternative as a source of embryonic stem cells, although many aspects related to the biology of parthenogenetic embryos and parthenogenetically derived cell lines still need to be elucidated. The present work was conducted to investigate the gene expression profile of rabbit parthenote embryos cultured under in vivo conditions using microarray analysis. Transcriptomic profiles indicate 2541 differentially expressed genes between parthenotes and normal in vivo fertilised blastocysts, of which 76 genes were upregulated and 16 genes downregulated in in vivo cultured parthenote blastocyst, using 3 fold-changes as a cut-off. While differentially upregulated expressed genes are related to transport and protein metabolic process, downregulated expressed genes are related to DNA and RNA binding. Using microarray data, 6 imprinted genes were identified as conserved among rabbits, humans and mice: GRB10, ATP10A, ZNF215, NDN, IMPACT and SFMBT2. We also found that 26 putative genes have at least one member of that gene family imprinted in other species. These data strengthen the view that a large fraction of genes is differentially expressed between parthenogenetic and normal embryos cultured under the same conditions and offer a new approach to the identification of imprinted genes in rabbit.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Bionic Buffer, 10X Concentrate, for Northern and Southern blotting, solution