Ugrás a tartalomra
Merck
Solid-state 207Pb NMR studies of mixed lead halides, PbFX (X=Cl, Br, or I).
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-07-29)
Alicia Glatfelter, Cecil Dybowski, David D Kragten, Shi Bai, Dale L Perry, Jenny Lockard
KIVONAT
Solid-state 207Pb NMR studies have been conducted on mixed lead(II) halides of the type PbFX, where X=Cl, Br, or I. NMR data for the mixed halides are compared to the solid-state NMR data for the divalent, binary lead halides, PbX2 (X=F, Cl, Br, I). The NMR data are evaluated in the context of the structures of the compounds and the effects of the mixed halides on the electronic structure of the divalent lead. Data sets for the mixed halides are discussed and compared to those for the regular lead(II) halides.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Lead(II) fluoride, 99.99% trace metals basis
Sigma-Aldrich
Lead(II) fluoride, powder, ≥99%