Ugrás a tartalomra
Merck
  • Effect of surface modification on semiconductor nanocrystal fluorescence lifetime.
Effect of surface modification on semiconductor nanocrystal fluorescence lifetime.
Chemphyschem : a European journal of chemical physics and physical chemistry (2011-03-03)
Maria J Ruedas-Rama, Angel Orte, Elizabeth A H Hall, Jose M Alvarez-Pez, Eva M Talavera
KIVONAT
Semiconductor nanocrystals, namely, quantum dots (QDs), present a set of unique photoluminescence properties, which has led to increased interest in using them as advantageous alternatives to conventional organic dyes. Many applications of QDs involve surface modification to enhance the solubility or biocompatibility of the QDs. One of the least exploited properties of QDs is the very long photoluminescence lifetime that usually has complex kinetics owing to the effect of quantum confinement. Herein, we describe the effect of different surface modifications on the photoluminescence decay kinetics of QDs. The different surface modifications were carefully chosen to provide lipophilic or water-soluble QDs with either positive or negative surface net charges. We also survey the effect on the QD lifetime of several ligands that interact with the QD surface, such as organic chromophores or fluorescent proteins. The results obtained demonstrate that time-resolved fluorescence is a useful tool for QD-based sensing to set the basis for the development of time-resolved-based nanosensors.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Hexadecylamine, 98%
Sigma-Aldrich
Hexadecylamine, technical grade, 90%