Ugrás a tartalomra
Merck
  • Urinary organic acid screening in children with developmental language delay.
Urinary organic acid screening in children with developmental language delay.
Journal of inherited metabolic disease (1999-10-13)
M Michelson, S Harel, A Gutman, T Lerman-Sagie
KIVONAT
The prevalence of 3-methylglutaconic aciduria was evaluated among children with developmental language disorders. A urine specimen was obtained from 40 children referred for developmental language delay to the Tel-Aviv Child Development Center during 12/96-6/97 and from 50 age-matched controls. Urine organic acids were analysed by gas chromatography-mass spectrometry. Urinary 3-methylglutaconic acid was quantified. A mildly increased excretion of 3-methylglutaconic acid was found in 8 children with developmental language delay. The combined excretion of 3-methylglutaconic and 3-methylglutaric acid was increased in 9 patients. There were no differences in the excretion of other organic acids. The patients with elevated 3-methylglutaconic acid did not differ from the other patients with developmental language disorders in any of the parameters evaluated. Mildly elevated urinary levels of 3-methylglutaconic acid may be a marker of a still undefined metabolic disorder presenting with developmental language delay. A further study in large groups of children with different developmental disorders is mandatory.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
3-Methylglutaric acid, 99%