Ugrás a tartalomra
Merck
  • Enzymatic activity and thermal stability of metallo proteins in hydrated ionic liquids.
Enzymatic activity and thermal stability of metallo proteins in hydrated ionic liquids.
Biopolymers (2010-07-29)
Kyoko Fujita, Hiroyuki Ohno
KIVONAT
Hydrated choline dihydrogen phosphate (Hy[ch][dhp]) containing 30 wt% water was investigated as a novel protein solvent. The Hy[ch][dhp] dissolved some metallo proteins (cytochrome c, peroxidase, ascorbate oxidase, azurin, pseudoazurin and fructose dehydrogenase) without any modification. These proteins retained the surroundings of the active site after dissolution in Hy[ch][dhp]. Some metallo proteins were found to retain their activity in the Hy[ch][dhp].
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Ascorbate Oxidase from Cucurbita sp., lyophilized powder, 1,000-3,000 units/mg protein