Ugrás a tartalomra
Merck
  • Oligogalacturonide-accelerated healing of mechanical wounding in tomato fruit requires calcium-dependent systemic acquired resistance.
Oligogalacturonide-accelerated healing of mechanical wounding in tomato fruit requires calcium-dependent systemic acquired resistance.
Food chemistry (2020-09-14)
Laifeng Lu, Ying Yang, Haoran Zhang, Dandan Sun, Zhenjing Li, Qingbin Guo, Changlu Wang, Liping Qiao
KIVONAT
Mechanical wounding causes significant economic losses of fresh produce due to accelerated senescence and spoilage as well as loss of nutritional value. Here, pre-application of oligogalacturonides (OGs) enzymatically hydrolyzed from apple pectin effectively reduced the healing times of mechanical wounds from>24 h in mock groups to 12 h, and the Botrytis cinerea infection rate was reduced from 37.5% to 12.5%. OGs accordingly increased callose deposition; SlPR1, SlPAL and SlHCT gene expression; and phenylalanine ammonia-lyase (PAL) activity around the wounds. Inhibition of Ca2+ signaling using the inhibitor Ruthenium Red markedly inhibited OG accelerated healing of mechanical wounding on fruit. SlPG2, SlEXP1, and SlCEL2 mRNAs accumulation was reduced in OG-elicited tomato fruit compared to water-treated fruit with subsequent retardation of the fruit softening during ripening. These results indicated that apple pectin OGs accelerate wound healing and inhibit fruit softening by activating calcium signaling in tomato fruits during postharvest storage.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Pectin from apple