Ugrás a tartalomra
Merck
  • UHPLC/ESI Q-Orbitrap Quantitation of 655 Pesticide Residues in Fruits and Vegetables-A Companion to an nDATA Working Flow.
UHPLC/ESI Q-Orbitrap Quantitation of 655 Pesticide Residues in Fruits and Vegetables-A Companion to an nDATA Working Flow.
Journal of AOAC International (2020-11-05)
Jian Wang, Daniel Leung, Willis Chow, Jon W Wong, James Chang
KIVONAT
Effective and expansive methods for multiresidue pesticide analysis are desired for routine monitoring programs. These methods are complex, especially when several hundred pesticides are involved. Two approaches to sort data and identify isomers and isobaric ions in pesticide mixtures were evaluated to determine whether they could be differentiated by mass resolving power and/or chromatographic resolution. This study presents an application of ultra-high performance liquid chromatography electrospray Q-Orbitrap mass spectrometry (UHPLC/ESI Q-Orbitrap) along with QuEChERS for the quantitation of 655 pesticide residues in fruits and vegetables. From the developed method, 94.7% of the 655 pesticides in fruits and 93.9% of those in vegetables had recoveries between 81% and 110%; 98.3% in both fruits and vegetables had an intermediate precision of ≤20%; and 97.7% in fruits or 97.4% in vegetables showed measurement uncertainty of ≤50%. When the retention time difference (ΔtR) of two isomers was ≥0.12 min, they were chromatographically resolved. Twenty five out of 35 pairs or groups of isomers were chromatographically separated (ΔtR ≥ 0.12 min), but 14 pairs were not resolved (ΔtR < 0.12 min). There were 493 pairs of pesticides with a mass-to-charge difference of <1 Da. Only one pair of isobaric ions could not be separated by mass and chromatographic resolution. UHPLC/ESI Q-Orbitrap along with QuEChERS sample preparation offers a practical quantitative companion method to a non-target data acquisition for target analysis workflow for pesticide residue analysis in routine monitoring programs for food safety.
ANYAGOK
Cikkszám
Márka
Termékleírás
Supelco
Carbofuran-d3, PESTANAL®, analytical standard