Ugrás a tartalomra
Merck
Lupane triterpenoids from Maytenus species.
Journal of natural products (2005-07-26)
Marvin J Núñez, Carolina P Reyes, Ignacio A Jiménez, Laila Moujir, Isabel L Bazzocchi
KIVONAT
Five new lupane triterpenes (1-5), in addition to 24 known ones, were isolated from Maytenus cuzcoina and M. chiapensis. Their structures were elucidated on the basis of spectroscopic analysis, including homonuclear and heteronuclear correlation NMR (COSY, ROESY, HSQC, and HMBC) experiments. The compounds were assayed for antimicrobial and cytotoxic activities, with 3-epi-betulinic acid and 28,30-dihydroxy-3-oxolup-20(29)-ene showing moderate cytotoxicity.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Betulin, ≥98%
Sigma-Aldrich
Lupeol, ≥94%