Ugrás a tartalomra
Merck
Gene yerP, involved in surfactin self-resistance in Bacillus subtilis.
Antimicrobial agents and chemotherapy (2001-11-16)
K Tsuge, Y Ohata, M Shoda
KIVONAT
Surfactin is a cyclic lipopeptide biosurfactant. Transposon mutagenesis was performed in Bacillus subtilis strain 168, and a surfactin-susceptible mutant, strain 801, was isolated. Analysis of the region of insertion revealed that yerP was the determinant of surfactin self-resistance. YerP had homology with the resistance, nodulation, and cell division (RND) family proton motive force-dependent efflux pumps only characterized in gram-negative strains. The yerP-deficient strain 802, in which the internal region of the yerP gene of B. subtilis strain 168 was deleted, showed susceptibility to acriflavine and ethidium bromide. When strain 802 was converted to a surfactin producer by introducing a functional sfp which encodes a 4'-phosphopantetheinyl transferase and is mutated in B. subtilis strain 168, this yerP-deficient strain produced surfactin, although surfactin production was significantly reduced. The expression of yerP was at its maximum at the end of the logarithmic growth phase and was not induced by surfactin. yerP is the first RND-like gene characterized in gram-positive strains and is supposed to be involved in the efflux of surfactin.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Surfactin, from Bacillus subtilis, ≥98.0% (HPLC)