Ugrás a tartalomra
Merck
  • Conversion of a decarboxylating to a non-decarboxylating glutaryl-coenzyme A dehydrogenase by site-directed mutagenesis.
Conversion of a decarboxylating to a non-decarboxylating glutaryl-coenzyme A dehydrogenase by site-directed mutagenesis.
FEBS letters (2011-04-12)
Jörg Schaarschmidt, Simon Wischgoll, Hans-Jörg Hofmann, Matthias Boll
KIVONAT
Glutaryl-coenzyme A (CoA) dehydrogenases (GDHs) are acyl-CoA dehydrogenases, which usually dehydrogenate and decarboxylate the substrate to crotonyl-CoA. In some anaerobic bacteria, non-decarboxylating GDHs exist that release glutaconyl-CoA (2,3-dehydroglutaryl-CoA) without decarboxylation. The differing mechanisms of decarboxylating and non-decarboxylating GDHs were investigated by site-directed mutagenesis of the gene coding for the crotonyl-CoA-forming GDH from Geobacter metallireducens. Exchange of single amino acids involved in substrate carboxylate binding impaired the decarboxylation step, resulting in relative glutaconyl-CoA:crotonyl-CoA formation rates of 1:1 (S97A) or 13:1 (Y370A). The total amount of glutaconyl-CoA formed was maximal in the Y370V+S97A double mutant. The results obtained indicate that an invariant deprotonated Tyr plays a crucial role for optimizing the leaving group potential of CO(2) in decarboxylating GDHs.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Glutaryl coenzyme A lithium salt, ≥90%