Ugrás a tartalomra
Merck
A mimetic peptide of α2,6-sialyllactose promotes neuritogenesis.
Neural regeneration research (2019-12-12)
Shuang-Xi Chen, Jia-Hui He, Yong-Jian Mi, Hui-Fan Shen, Melitta Schachner, Wei-Jiang Zhao
KIVONAT
Oxidative stress contributes to the pathogenesis of neurodegenerative diseases. With the aim to find reagents that reduce oxidative stress, a phage display library was screened for peptides mimicking α2,6-sialyllactose (6'-SL), which is known to beneficially influence neural functions. Using Sambucus nigra lectin, which specifically binds to 6'-SL, we screened a phage display library and found a peptide comprising identical sequences of 12 amino acids. Mimetic peptide, reverse peptide and scrambled peptide were tested for inhibition of 6'-SL binding to the lectin. Indeed, lectin binding to 6'-SL was inhibited by the most frequently identified mimetic peptide, but not by the reverse or scrambled peptides, showing that this peptide mimics 6'-SL. Functionally, mimetic peptide, but not the reverse or scrambled peptides, increased viability and expression of neural cell adhesion molecule L1 in SK-N-SH human neuroblastoma cells, and promoted survival and neurite outgrowth of cultured mouse cerebellar granule neurons challenged by H2O2-induced oxidative stress. The combined results indicate that the 6'-SL mimetic peptide promotes neuronal survival and neuritogenesis, thus raising hopes for the treatment of neurodegenerative diseases. This study was approved by the Medical Ethics Committee of Shantou University Medical College, China (approval No. SUMC 2014-004) on February 20, 2014.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
6′-Sialyllactose sodium salt, from bovine milk or colostrum, ≥97% (HPLC)