Ugrás a tartalomra
Merck
  • Phosphorylation and ubiquitination of the transcription factor sterol regulatory element-binding protein-1 in response to DNA binding.
Phosphorylation and ubiquitination of the transcription factor sterol regulatory element-binding protein-1 in response to DNA binding.
The Journal of biological chemistry (2006-07-11)
Tanel Punga, Maria T Bengoechea-Alonso, Johan Ericsson
KIVONAT
Members of the sterol regulatory element-binding protein (SREBP) family of transcription factors control cholesterol and lipid metabolism and play critical roles during adipocyte differentiation. The transcription factor SREBP1 is degraded by the ubiquitin-proteasome system following phosphorylation of Thr426 and Ser430 in its phosphodegron. We now demonstrate that the glycogen synthase kinase (GSK)-3beta-dependent phosphorylation of these residues in SREBP1 is enhanced in response to specific DNA binding. DNA binding enhances the direct interaction between the C-terminal domain of SREBP1 and GSK-3beta. Accordingly, we demonstrate that GSK-3beta is recruited to the promoters of SREBP target genes in vivo. As a result of the phosphorylation of Thr426 and Ser430, the ubiquitin ligase Fbw7 is recruited to SREBP molecules associated with target promoters. Using a reconstituted ubiquitination system, we demonstrate that Fbw7-mediated ubiquitination of SREBP1 is dependent on its DNA binding activity. Thus, DNA binding could provide a mechanistic link between the phosphorylation, ubiquitination, and degradation of active transcription factors.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Anti-SREBP1 Antibody, clone 2121, clone 2121, Upstate®, from mouse