Ugrás a tartalomra
Merck
  • Liver X receptor antagonist reduces lipid formation and increases glucose metabolism in myotubes from lean, obese and type 2 diabetic individuals.
Liver X receptor antagonist reduces lipid formation and increases glucose metabolism in myotubes from lean, obese and type 2 diabetic individuals.
Diabetologia (2007-07-31)
E T Kase, G H Thoresen, S Westerlund, K Højlund, A C Rustan, M Gaster
KIVONAT
Liver X receptors (LXRs) play important roles in lipid and carbohydrate metabolism. The purpose of the present study was to evaluate effects of the endogenous LXR agonist 22-R-hydroxycholesterol (22-R-HC) and its stereoisomer 22-S-hydroxycholesterol (22-S-HC), in comparison with the synthetic agonist T0901317 on lipid and glucose metabolism in human skeletal muscle cells (myotubes). Myotubes established from lean and obese control volunteers and from obese type 2 diabetic volunteers were treated with LXR ligands for 4 days. Lipid and glucose metabolisms were studied with labelled precursors, and gene expression was analysed using real-time PCR. Treatment with T0901317 increased lipogenesis (de novo lipid synthesis) and lipid accumulation in myotubes, this increase being more pronounced in myotubes from type 2 diabetic volunteers than from lean volunteers. Furthermore, 22-S-HC efficiently counteracted the T0901317-induced enhancement of lipid formation. Moreover, synthesis of diacylglycerol, cholesteryl ester and free cholesterol from acetate was reduced below baseline by 22-S-HC, whereas glucose uptake and oxidation were increased. Both 22-S-HC and 22-R-HC, in contrast to T0901317, decreased the expression of genes involved in cholesterol synthesis, whereas only 22-R-HC, like T0901317, increased the expression of the gene encoding the reverse cholesterol transporter ATP-binding cassette subfamily A1 (ABCA1). T0901317-induced lipogenesis and lipid formation was more pronounced in myotubes from type 2 diabetic patients than from lean individuals. 22-S-HC counteracted these effects and reduced de novo lipogenesis below baseline, while glucose uptake and oxidation were increased.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
22(S)-Hydroxycholesterol