Ugrás a tartalomra
Merck
  • Preparation of the Ru3(CO)8-pyridine-alcohol cluster and its use for the selective catalytic transformation of primary to secondary amines.
Preparation of the Ru3(CO)8-pyridine-alcohol cluster and its use for the selective catalytic transformation of primary to secondary amines.
Dalton transactions (Cambridge, England : 2003) (2018-09-21)
Ajeet Singh, Shaikh M Mobin, Pradeep Mathur
KIVONAT
The synthesis of pyridine alcohol based ruthenium carbonyl clusters Ru3(hep)2(CO)8 (1), Ru3(hpp)2(CO)8 (2), and Ru3(bhmp-H)2(CO)8 (3) {hep-H = 2-(2-hydroxyethyl)pyridine, hpp-H = 2-(3-hydroxypropyl)pyridine and bhmp-H2 = 2,6-bis(hydroxymethyl)pyridine} has been carried out by the reaction of the corresponding pyridine-alcohol ligands with Ru3(CO)12. Clusters 1-3 have been characterized using elemental analysis, NMR, FT-IR, mass spectrometry and single-crystal X-ray structures. The clusters were explored for the selective catalytic transformation of primary amines into secondary amines using alcohols as the mono-alkylating agents via hydrogen transfer reactions. All three display efficient catalytic activity with 1 being the most effective.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Triruthenium dodecacarbonyl, 99%