Ugrás a tartalomra
Merck
  • Palladium-Catalyzed Arylation of Arylboronic Acids with Yagupolskii-Umemoto Reagents.
Palladium-Catalyzed Arylation of Arylboronic Acids with Yagupolskii-Umemoto Reagents.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-03-08)
Shi-Meng Wang, Xiao-Yan Wang, Hua-Li Qin, Cheng-Pan Zhang
KIVONAT
A Pd-catalyzed Suzuki cross-coupling of arylboronic acids with Yagupolskii-Umemoto reagents was explored. In contrary to trifluoromethylations, the Pd-catalyzed reaction of R-B(OH)2 and [Ar2 SCF3 ](+) [OTf](-) provided the arylation products (R-Ar) in good to high yields. The reaction confirms that the S-Ar bonds of [Ar2 SCF3 ](+) [OTf](-) can be readily cleaved in the presence of Pd complexes. The relatively electron-poor aryl groups of asymmetric [Ar(1) Ar(2) SCF3 ](+) [OTf](-) salts are more favorably transferred compared to the electron-rich ones. This reaction represents the first report of utilization of [Ar2 SCF3 ](+) [OTf](-) as arylation reagents in organic synthesis.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Mes-Umemoto reagent, ≥95%