Fontos dokumentumok
Z422932
Sigma-Aldrich
Aldrich® pressure filter/drying funnel assemblies with removable fritted disc
funnel capacity 2,000 mL
About This Item
UNSPSC kód:
41121809
NACRES:
NB.24
pórusméret:
coarse
tölcsérkapacitás:
2,000 mL
material:
PTFE (Bottom support)
gyártó/kereskedő neve:
Aldrich
material
PTFE (Bottom support)
gyártó/kereskedő neve
Aldrich
tölcsérkapacitás
2,000 mL
pórusméret
coarse
kapacitás
2000 mL capacity
Általános leírás
Pressure or vacuum can be applied alternately or simultaneously for fast filtration or drying. Unique design allows the fritted disc to be removed and replaced. The bottom support is all PTFE and has a vent. A port is supplied on the top cap for an optional adjustable pressure relief valve. Large top and bottom openings are 85 mm I.D. allowing easy access to funnel contents. Complete assembly includes top cap, funnel, coarse frit, 90 mm base assembly, and 2 L bottle with GL 45 thread.
Jogi információk
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Válasszon a legfrissebb verziók közül:
Analitikai tanúsítványok (COA)