主要文件
51272-U
MicroTight 转接头
PEEK, for use with (1/16" to 1/32" tubing)
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115700
NACRES:
SB.52
物料
PEEK
相容性
for use with (1/16" to 1/32" tubing)
正在寻找类似产品? 访问 产品对比指南
一般描述
These adapters will help create a true zero dead volume (ZDV) connection between 1/16" I.D. tubing and capillary tubing. MicroTight Adapters allow connection of 1/16" O.D. tubing directly to 1/32" O.D. and 360 μm tubing.
推荐产品
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
历史批次信息供参考:
分析证书(COA)