33136
1 of 10
Supelco
27312
需要 8mm 密封盖的钳口微量样品瓶
SU860015
Crimp seals (magnetic) with PTFE/silicone septa, pkg/100
508888
透明的 1.2mL 钳口平底样品瓶,8×40mm
27454-U
带 PTFE/橡胶隔垫的钳口密封盖
27362
带 PTFE/硅胶隔垫的钳口密封盖
33233-U
Sigma-Aldrich
SMB00472
氯化帕马汀
33283
钳口样品瓶启盖器
27452
H8502
盐酸多巴胺 盐酸盐
PTFE/red rubber septumsilver aluminum seal
open center
pkg of 1000 ea
8 mm
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.