Documentos clave
Z100307
Sigma-Aldrich
Aldrich® West condenser w/ Clear-Seal joint
joint: ST/NS 19/22, pkg of 1 ea
Seleccione un Tamaño
1 EA
127,00 €
127,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Seleccione un Tamaño
Cambiar Vistas
1 EA
127,00 €
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
127,00 €
Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad
Junta
joint: ST/NS19/22
envase
pkg of 1 ea
Longitud
200 mm
Descripción general
Replacement part for the Diazald Kit, Z100250.
The Diazald kit is a set of distillation glassware designed for the safe preparation of diazomethane (~100 mmol). Glassware without sharp edges or ground-glass joints are recommended by de Boer and Backer [Org. Syn., Coll. Vol. 4, 250 (1963)]. The Diazald kit features 19/22 Clear-Seal joints; these joints do not require grease even for vacuum applications, thus avoiding a source of contamination.
The glassware in the kit should be washed with care and without the use of wire brushes which will scratch the inner surface. For further information, request Technical Information Bulletin No. AL-180 and for a review on the preparation and reactions of diazomethane, see Aldrichim. Acta, 16, 1, 1983.
Call or visit our website for pricing on replacement parts.
NOTE: We are unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)