Merck
Z763470
MAGic Clamp universal platform for flasks and tube racks
L × W 9.5 in. × 11.5 in.
Synonym(e):
magnetic clamps, orbital shaker platforms
Größe auswählen
1 EA
319,00 €
319,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
319,00 €
About This Item
UNSPSC-Code:
41103811
NACRES:
NB.77
319,00 €
Check Cart for Availability
Hersteller/Markenname
Benchmark Scientific H1000-MR
L × B
9.5 in. × 11.5 in.
Allgemeine Beschreibung
The convenient MAGic Clamp platforms (large or small) are available for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is widely recognized as the easiest way to instantly change between flask clamps of different sizes.
Rechtliche Hinweise
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)